1-(Methylsulfonyl)-4-nitrobenzene 97%

Warning

B20068-03 B20068-03 B20068-06 B20068-06
AAB20068-03EA 53.94 USD
CAAAB20068-06 AAB20068-03 CAAAB20068-03 AAB20068-06
1-(Methylsulfonyl)-4-nitrobenzene 97%
1-(Methylsulphonyl)-4-nitrobenzene
Formula: C₇H₇NO₄S
Melting Pt: 136…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00047803
CAS Number: 2976-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR