2,4,6-Trimethyl-4-heptanol 97%
|
Warning
|
B20059-06
B20059-06
B20059-03
B20059-03
B20059-14
B20059-14
AAB20059-06EA
86.26
USD
AAB20059-06
AAB20059-03
AAB20059-14
CAAAB20059-06
CAAAB20059-03
CAAAB20059-14
2,4,6-Trimethyl-4-heptanol 97%
2,4,6-Trimethyl-4-heptanol
Formula:
C₁₀H₂₂O Storage Temperature: Ambient |
MDL Number:
MFCD00048354 CAS Number: 60836-07-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...