2-Hepten-4-ol mixture E/Z 98%

Warning

B20057-14 B20057-14 B20057-06 B20057-06 B20057-03 B20057-03
AAB20057-14EA 368.74 USD
AAB20057-14 AAB20057-03 CAAAB20057-14 CAAAB20057-03 CAAAB20057-06 AAB20057-06
2-Hepten-4-ol mixture E/Z 98%
2-Hepten-4-ol
Formula: C₇H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD00021932
CAS Number: 4798-59-8
UN: 1987
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR