4-Bromo-2-ethyl-1-iodobenzene 97% stabilized

Supplier: Thermo Scientific Chemicals

Warning


Stabilization: Stabilised with copper

B20052-09 B20052-88 B20052-88 B20052-09
AAB20052-09EA 177 USD
CAAAB20052-88 AAB20052-09 CAAAB20052-09 AAB20052-88
4-Bromo-2-ethyl-1-iodobenzene 97% stabilized
4-Bromo-2-ethyl-1-iodobenzene
Formula: C₈H₈BrI
MW: 310.97 g/mol
Boiling Pt: 84…86 °C/0.4 mm
Storage Temperature: Ambient
MDL Number: MFCD00221458
CAS Number: 175278-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR