Methyl-5-chlorosalicylate 97%

Warning

Synonyms: Methyl 5-chloro-2-hydroxybenzoate

A19971-88 A19971-09 A19971-88 A19971-09
AAA19971-88EA 70.11 USD
CAAAA19971-09 AAA19971-88 CAAAA19971-88 AAA19971-09
Methyl-5-chlorosalicylate 97%
Methyl 5-chlorosalicylate
Formula: C₈H₇ClO₃
MW: 186.60 g/mol
Melting Pt: 44…48 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00016461
CAS Number: 4068-78-4
EINECS: 223-778-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR