4'-Bromo-3'-(trifluoromethyl)acetanilide 97%

Warning

Synonyms: N-(4-Bromo-3-(trifluoromethyl)phenyl)acetamide

A19939-09 A19939-09 A19939-88 A19939-88
AAA19939-09EA 157.59 USD
AAA19939-09 CAAAA19939-09 AAA19939-88 CAAAA19939-88
4'-Bromo-3'-(trifluoromethyl)acetanilide 97%
4'-Bromo-3'-(trifluoromethyl)acetanilide
Formula: C₉H₇BrF₃NO
MW: 282.07 g/mol
Melting Pt: 116…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00052361
CAS Number: 41513-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR