4-Ethyl-3-hexanol 95%

Warning

A19924-03 A19924-06 A19924-14 A19924-03 A19924-14 A19924-06
AAA19924-06EA 144.3 USD
AAA19924-06 CAAAA19924-06 CAAAA19924-03 CAAAA19924-14 AAA19924-14 AAA19924-03
4-Ethyl-3-hexanol 95%
4-Ethyl-3-hexanol
Formula: C₈H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD00021917
CAS Number: 19780-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR