1,5,9-Decatriene cis- and trans- mixture 97%

Warning

A19822-AA A19822-AA A19822-AC A19822-AC
AAA19822-AAEA 55.05 USD
CAAAA19822-AA CAAAA19822-AC AAA19822-AA AAA19822-AC
1,5,9-Decatriene cis- and trans- mixture 97%
1,5,9-Decatriene
Formula: C₁₀H₁₆
Boiling Pt: 54…55 °C/14 mm
Density: 0.765
Flash Pt: 46 °C(115 °F)
Storage Temperature: Ambient
MDL Number: MFCD00055625
CAS Number: 13393-64-1
EINECS: 236-474-2
UN: 3295
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR