Methylphenylphosphinic acid 98%

Warning

A19774-06 A19774-14 A19774-03
CAAAA19774-14 CAAAA19774-03 CAAAA19774-06
Methylphenylphosphinic acid 98%
Methylphenylphosphinic acid
Formula: C₇H₉O₂P
Melting Pt: 132…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00123313
CAS Number: 4271-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR