3',5'-Dibenzyloxyacetophenone 98%

Synonyms: 1-[3,5-Bis(benzyloxy)phenyl]ethan-1-one

A19614-09 A19614-30 A19614-18 A19614-09 A19614-30 A19614-18
AAA19614-18EA 116.1 USD
AAA19614-18 CAAAA19614-09 AAA19614-09 CAAAA19614-30 CAAAA19614-18 AAA19614-30
3',5'-Dibenzyloxyacetophenone 98%
3',5'-Dibenzyloxyacetophenone
Formula: C₂₂H₂₀O₃
MW: 332.40
Melting Pt: 60…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00004777
CAS Number: 28924-21-2
EINECS: 249-315-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR