2-Amino-4-phenylthiazole hydrobromide monohydrate 99%

Warning

A19542-06 A19542-06 A19542-22 A19542-14 A19542-22 A19542-14
AAA19542-06EA 35.98 USD
AAA19542-06 CAAAA19542-06 CAAAA19542-14 AAA19542-22 CAAAA19542-22 AAA19542-14
2-Amino-4-phenylthiazole hydrobromide monohydrate 99%
2-Amino-4-phenylthiazole hydrobromide monohydrate
Formula: C₉H₈N₂S.HBr.H₂O
MW: 275.17 g/mol (257.15 anhy)
Melting Pt: 180…183 °C
Storage Temperature: Ambient
MDL Number: MFCD00150083
CAS Number: 52253-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR