2-(Isothiocyanatomethyl)furan 97%

Warning

A19410-06 A19410-06 A19410-14 A19410-14
AAA19410-06EA 69.2 USD
AAA19410-06 CAAAA19410-06 CAAAA19410-14 AAA19410-14
2-(Isothiocyanatomethyl)furan 97%
2-(Isothiocyanatomethyl)furan
Formula: C₆H₅NOS
Boiling Pt: 64…68 °C/2 mm
Density: 1.390
Storage Temperature: Refrigerator
MDL Number: MFCD00014112
CAS Number: 4650-60-6
EINECS: 225-079-0
UN: 2922
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR