3-Bromo-4-fluorobenzylamine hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A19280-06
A19280-06
A19280-14
A19280-14
AAA19280-06EA
82.49
USD
AAA19280-06
CAAAA19280-06
CAAAA19280-14
AAA19280-14
3-Bromo-4-fluorobenzylamine hydrochloride 98%
3-Bromo-4-fluorobenzylamine hydrochloride
Formula:
C₇H₈BrClFN MW: 240.50 g/mol Melting Pt: 214…218 °C Storage Temperature: Ambient |
MDL Number:
MFCD00143094 CAS Number: 202865-68-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...