4-Butylbenzonitrile 98%

Warning

A19174-22 A19174-14 A19174-14 A19174-06 A19174-22 A19174-06
AAA19174-22EA 859.14 USD
CAAAA19174-06 AAA19174-22 CAAAA19174-14 AAA19174-14 CAAAA19174-22 AAA19174-06
4-Butylbenzonitrile 98%
4-Butylbenzonitrile
Formula: C₁₁H₁₃N
MW: 159.23 g/mol
Boiling Pt: 130…132 °C/15 mm
Density: 0.924
Storage Temperature: Ambient
MDL Number: MFCD00052345
CAS Number: 20651-73-4
UN: 3276
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR