4-n-Butyl-2-methylphenyl isothiocyanate 95%

Danger

A18984-06 A18984-03 A18984-03 A18984-06 A18984-14 A18984-14
AAA18984-06EA 88.54 USD
CAAAA18984-03 CAAAA18984-14 AAA18984-06 AAA18984-03 AAA18984-14 CAAAA18984-06
4-n-Butyl-2-methylphenyl isothiocyanate 95%
4-n-Butyl-2-methylphenyl isothiocyanate
Formula: C₁₂H₁₅NS
MW: 205.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00276966
CAS Number: 175205-37-5
UN: 2922
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR