4-Hydroxyphenylacetone 98+%

Warning

A18936-14 A18936-06 A18936-14 A18936-06
AAA18936-14EA 389.84 USD
CAAAA18936-14 AAA18936-14 AAA18936-06 CAAAA18936-06
4-Hydroxyphenylacetone 98+%
4-Hydroxyphenylacetone
Formula: C₉H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00210439
CAS Number: 770-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR