Ethyl-3-methylvalerate 98%

Warning

Synonyms: 3-Methylvaleric Acid Ethyl EsterEthyl 3-Methylpentanoate3-Methylpentanoic Acid Ethyl Ester

A18868-06 A18868-06 A18868-14 A18868-14
AAA18868-06EA 35.06 USD
AAA18868-06 CAAAA18868-06 CAAAA18868-14 AAA18868-14
Ethyl-3-methylvalerate 98%
Ethyl-3-methylvalerate
Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 158 °C
Density: 0.87
Flash Pt: 45 °C(113 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041390
CAS Number: 5870-68-8
EINECS: 227-524-4
UN: 3272
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR