4-Ethylphenylacetone 97+%

Supplier: Thermo Scientific Chemicals

A18810-03 A18810-14 A18810-06 A18810-14 A18810-06
AAA18810-06EA 159 USD
AAA18810-06 AAA18810-03 AAA18810-14 CAAAA18810-14 CAAAA18810-06
4-Ethylphenylacetone 97+%
4-Ethylphenylacetone
Formula: C₁₁H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD00210429
CAS Number: 75251-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR