2-Amino-1-naphthalenesulfonic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Aminonaphthalene-1-sulfonic acid2-Aminonaphthalene-1-sulphonic acid

A18644-36 A18644-36 A18644-0E A18644-0E
AAA18644-36EA 71.09 USD
CAAAA18644-0E CAAAA18644-36 AAA18644-36 AAA18644-0E
2-Amino-1-naphthalenesulfonic acid 98%
2-Amino-1-naphthalenesulphonic acid
Formula: C₁₀H₉NO₃S
MW: 223.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003988
CAS Number: 81-16-3
EINECS: 201-331-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR