2-Acetamidoethanol 90+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-Acetylethanolamine

A18638-22 A18638-36 A18638-22 A18638-36
AAAA18638-36EA 112.27 USD
CAAAAA18638-22 AAAA18638-36 CAAAAA18638-36 AAAA18638-22
2-Acetamidoethanol 90+%
2-Acetamidoethanol
Formula: C₄H₉NO₂
MW: 103.12 g/mol
Boiling Pt: 151…155 °C/5 mm
Density: 1.120
Flash Pt: 177 °C(351 °F)
Storage Temperature: Ambient
EINECS: 205-530-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR