8-Octanelactam 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Azacyclononanone
, 8-Octanolactam
A18621-22
A18621-22
A18621-06
A18621-14
A18621-14
A18621-06
AAA18621-14EA
350.99
USD
CAAAA18621-22
CAAAA18621-14
AAA18621-14
CAAAA18621-06
AAA18621-22
AAA18621-06
8-Octanelactam 98%
8-Octanelactam
Formula:
C₈H₁₅NO MW: 141.21 g/mol Melting Pt: 72…75 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003273 CAS Number: 935-30-8 EINECS: 213-300-3 UN: 2811 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...