2-(2-Naphthoyl)-1-(p-toluoyl)hydrazine 98%

Warning

A18590-03 A18590-06 A18590-06 A18590-03
AAA18590-03EA 32.75 USD
CAAAA18590-03 AAA18590-03 AAA18590-06 CAAAA18590-06
2-(2-Naphthoyl)-1-(p-toluoyl)hydrazine 98%
2-(2-Naphthoyl)-1-(p-toluoyl)hydrazine
Formula: C₁₉H₁₆N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00021629
CAS Number: 83803-95-6
EINECS: 280-916-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR