4-Benzoyl-4'-bromobiphenyl 99%

Supplier: Thermo Scientific Chemicals

A18482-22 A18482-14 A18482-36 A18482-22 A18482-36 A18482-14
AAA18482-22EA 238.99 USD
CAAAA18482-14 AAA18482-22 CAAAA18482-22 CAAAAA18482-36 AAAA18482-36 AAA18482-14
4-Benzoyl-4'-bromobiphenyl 99%
4-Benzoyl-4'-bromobiphenyl
Formula: C₁₉H₁₃BrO
Melting Pt: 155…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00209630
CAS Number: 63242-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR