2-Chloro-2-oxo-1,3,2-dioxaphospholane 95%

Danger

Synonyms: Ethylene glycol chlorophosphate2-Chloro-1,3,2-dioxaphospholane-2-oxide

A18320-06 A18320-14 A18320-14 A18320-06 A18320-22 A18320-22
AAA18320-06EA 103.82 USD
AAA18320-06 CAAAA18320-22 CAAAA18320-14 AAA18320-22 AAA18320-14 CAAAA18320-06
2-Chloro-2-oxo-1,3,2-dioxaphospholane 95%
2-Chloro-2-oxo-1,3,2-dioxaphospholane
Formula: C₂H₄ClO₃P
MW: 142.47 g/mol
Boiling Pt: 98…99 °C/2 mm
Melting Pt: 12…14 °C
Density: 1.56
Flash Pt: >110 °C(230 °F)
Storage Temperature: Freezer
MDL Number: MFCD00043138
CAS Number: 6609-64-9
EINECS: 229-560-6
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR