2,2,5,5-Tetramethyl-3-pyrroline-3-carboxamide 99%
Supplier: Thermo Scientific Chemicals
A18294-06
A18294-06
A18294-03
A18294-03
AAA18294-03EA
51.69
USD
AAA18294-03
CAAAA18294-06
AAA18294-06
CAAAA18294-03
2,2,5,5-Tetramethyl-3-pyrroline-3-carboxamide 99%
2,2,5,5-Tetramethyl-3-pyrroline-3-carboxamide
Formula:
C₉H₁₆N₂O Melting Pt: 179…183 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041846 CAS Number: 19805-75-5 EINECS: 243-331-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...