Dibromofluorescein 95% (dye content)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4',5'-Dibromofluorescein

A18226-18 A18226-30 A18226-18 A18226-30
AAA18226-30EA 95.19 USD
CAAAA18226-30 AAA18226-30 CAAAA18226-18 AAA18226-18
Dibromofluorescein 95% (dye content)
Dibromofluorescein
Formula: C₂₀H₁₀Br₂O₅
MW: 490.10 g/mol
Melting Pt: 270…273 °C
Storage Temperature: Ambient
MDL Number: MFCD00005042
CAS Number: 596-03-2
EINECS: 209-876-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR