N-(2-Cyanoethyl)glycine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-(2-Cyanoethyl)aminoacetic Acid

A18199-06 A18199-14 A18199-14 A18199-06
AAA18199-06EA 33.69 USD
CAAAA18199-06 AAA18199-06 CAAAA18199-14 AAA18199-14
N-(2-Cyanoethyl)glycine 98%
N-(2-Cyanoethyl)glycine
Formula: C₅H₈N₂O₂
MW: 128.13 g/mol
Melting Pt: ca 195 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00066903
CAS Number: 3088-42-4
EINECS: 221-418-1
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR