1,4-Diacetoxy-2-butyne 97%

Synonyms: Bis(acetoxymethyl)acetylene2-Butyne-1,4-diol Diacetate

A18182-14 A18182-14 A18182-06 A18182-06
AAA18182-06EA 49.71 USD
CAAAA18182-14 AAA18182-06 AAA18182-14 CAAAA18182-06
1,4-Diacetoxy-2-butyne 97%
1,4-Diacetoxy-2-butyne
Formula: C₈H₁₀O₄
MW: 170.16 g/mol
Boiling Pt: 116…118 °C/8 mm
Melting Pt: 27…28 °C
Density: 1.125
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014982
CAS Number: 1573-17-7
EINECS: 216-394-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR