5-Bromonicotinamide 98%

Warning

Synonyms: 5-Bromo-3-pyridinecarboxamide

A17840-14 A17840-06 A17840-14 A17840-06
AAA17840-14EA 112.78 USD
CAAAA17840-06 AAA17840-14 CAAAA17840-14 AAA17840-06
5-Bromonicotinamide 98%
5-Bromonicotinamide
Formula: C₆H₅BrN₂O
MW: 201.03 g/mol
Melting Pt: 226…227 °C
Storage Temperature: Ambient
MDL Number: MFCD00173919
CAS Number: 28733-43-9
EINECS: 244-065-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR