(Methoxycarbonylmethyl)triphenylphosphonium chloride 97%

Warning

A17775-30 A17775-18 A17775-09 A17775-18 A17775-09 A17775-30
AAAA17775-30EA 352.98 USD
CAAAAA17775-30 AAAA17775-30 AAA17775-18 AAA17775-09 CAAAA17775-09 CAAAA17775-18
(Methoxycarbonylmethyl)triphenylphosphonium chloride 97%
(Methoxycarbonylmethyl)triphenylphosphonium chloride
Formula: C₂₁H₂₀ClO₂P
MW: 370.82
Melting Pt: ca 162 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00050199
CAS Number: 2181-97-7
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR