(2,6-Dichlorobenzyl)triphenylphosphonium chloride 98%

Warning

A17651-22 A17651-22 A17651-06 A17651-14 A17651-14 A17651-06
AAA17651-06EA 60.87 USD
CAAAA17651-22 CAAAA17651-14 AAA17651-06 AAA17651-14 AAA17651-22 CAAAA17651-06
(2,6-Dichlorobenzyl)triphenylphosphonium chloride 98%
(2,6-Dichlorobenzyl)triphenylphosphonium chloride
Formula: C₂₅H₂₀Cl₃P
MW: 457.78 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00061392
CAS Number: 18583-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR