Dicinnamalacetone ≥97%

Supplier: Thermo Scientific Chemicals

Synonyms: 1,9-Diphenyl-1,3,6,8-nonatetraen-5-one

A17568-06 A17568-14 A17568-22 A17568-06 A17568-22 A17568-14
AAA17568-22EA 511.82 USD
CAAAA17568-14 CAAAA17568-06 AAA17568-22 AAA17568-06 AAA17568-14 CAAAA17568-22
Dicinnamalacetone ≥97%
Dicinnamalacetone
Formula: C₂₁H₁₈O
MW: 286.37 g/mol
Melting Pt: 141…143 °C
Storage Temperature: Ambient
MDL Number: MFCD00004792
CAS Number: 622-21-9
EINECS: 210-724-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR