1,2,3-Benzenetricarboxylic acid dihydrate 99%

Warning

Synonyms: 1,2,3-Benzenetricarboxylic acid hydrate

A17491-09 A17491-09 A17491-06 A17491-14 A17491-06 A17491-14
AAA17491-06EA 60.19 USD
CAAAA17491-06 AAA17491-06 AAA17491-14 CAAAAA17491-09 AAAA17491-09 CAAAA17491-14
1,2,3-Benzenetricarboxylic acid dihydrate 99%
1,2,3-Benzenetricarboxylic acid dihydrate
Formula: C₉H₆O₆·2H₂O
Melting Pt: ca 191 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149097
CAS Number: 36362-97-7
EINECS: 209-317-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR