4-Methoxystilbene 98%

Supplier: Thermo Scientific Chemicals
Warning

A17328-14 A17328-14 A17328-22 A17328-06 A17328-06 A17328-22
AAA17328-22EA 1580.44 USD
AAA17328-22 CAAAA17328-22 AAA17328-14 AAA17328-06 CAAAA17328-06 CAAAA17328-14
4-Methoxystilbene 98%
4-Methoxystilbene
Formula: C₁₅H₁₄O
MW: 210.27
Melting Pt: 136…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00017178
CAS Number: 1142-15-0
EINECS: 214-530-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR