2-(Benzimidazolylthio)acetic acid hydrazide 97%

Warning

Synonyms: 2-((1H-Benzo[d]imidazol-2-yl)thio)acetohydrazide

A17198-06 A17198-14 A17198-14 A17198-06
AAA17198-14EA 266.4 USD
CAAAA17198-14 CAAAA17198-06 AAA17198-14 AAA17198-06
2-(Benzimidazolylthio)acetic acid hydrazide 97%
2-(Benzimidazolylthio)acetic acid hydrazide
Formula: C₉H₁₀N₄O₄S
MW: 222.27 g/mol
Melting Pt: ca 160 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00171027
CAS Number: 30065-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR