Bis[4-(3-aminophenoxy)phenyl]sulfone 95%

Warning

Synonyms: 3,3'-((sulfonylbis(4,1-phenylene))bis(oxy))dianiline3,3'-((sulphonylbis(4,1-phenylene))bis(oxy))dianiline

A17194-0B A17194-18 A17194-0B A17194-18 A17194-30 A17194-30
AAA17194-30EA 178.73 USD
CAAAA17194-30 AAA17194-30 CAAAA17194-18 CAAAA17194-0B AAA17194-0B AAA17194-18
Bis[4-(3-aminophenoxy)phenyl]sulfone 95%
Bis[4-(3-aminophenoxy)phenyl]sulfone
Formula: C₂₄H₂₀N₂O₄S
MW: 432.49 g/mol
Melting Pt: 132…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00054738
CAS Number: 30203-11-3
EINECS: 250-091-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR