Holmium (III) sulfate hydrate 99%

Warning

A17092-14 A17092-14
AAAA17092-14EA 219.78 USD
AAAA17092-14 CAAAAA17092-14
Holmium (III) sulfate hydrate 99%
Holmium (III) sulphate hydrate
Formula: Ho₂O₁₂S₃·H₂O
MW: 618.03 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00149740
CAS Number: 15622-40-9
EINECS: 239-697-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR