1-[(2-Chloroethyl)sulfonyl]-4-fluorobenzene 98%

Warning

Synonyms: 2-Chloroethyl 4-fluorophenyl sulphone

A16930-03 A16930-03 A16930-06 A16930-06
AAA16930-06EA 149.85 USD
CAAAA16930-03 CAAAA16930-06 AAA16930-06 AAA16930-03
1-[(2-Chloroethyl)sulfonyl]-4-fluorobenzene 98%
1-[(2-Chloroethyl)sulphonyl]-4-fluorobenzene
Formula: C₈H₈ClFO₂S
MW: 222.67
Melting Pt: 67…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00203483
CAS Number: 33330-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR