2-Bromo-4,5-dimethoxycinnamic acid 97%

Warning

A16862-14 A16862-14 A16862-06 A16862-06 A16862-22 A16862-22
AAAA16862-22EA 445.11 USD
CAAAAA16862-22 CAAAA16862-06 CAAAA16862-14 AAAA16862-22 AAA16862-14 AAA16862-06
2-Bromo-4,5-dimethoxycinnamic acid 97%
2-Bromo-4,5-dimethoxycinnamic acid
Formula: C₁₀H₉BrO₃
Melting Pt: 253…256 °C
Storage Temperature: Ambient
MDL Number: MFCD00017614
CAS Number: 151539-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR