Methyl-2-acetamido-5-chlorobenzoate 97%

Synonyms: N-Acetyl-5-chloroanthranilic Acid Methyl Ester2-Acetamido-5-chlorobenzoic Acid Methyl EsterMethyl N-Acetyl-5-chloroanthranilate

A16826-30 A16826-18 A16826-30 A16826-18
AAA16826-18EA 51.94 USD
CAAAA16826-18 AAA16826-18 CAAAA16826-30 AAA16826-30
Methyl-2-acetamido-5-chlorobenzoate 97%
Methyl-2-acetamido-5-chlorobenzoate
Formula: C₁₀H₁₀ClNO₃
MW: 227.64 g/mol
Melting Pt: 128…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00144759
CAS Number: 20676-54-4
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR