Methyl quinoxaline-6-carboxylate 98%

Synonyms: Methyl 6-Quinoxalinecarboxylate

A16689-06 A16689-03 A16689-03 A16689-06
AAA16689-06EA 374.11 USD
AAA16689-06 CAAAA16689-06 AAA16689-03 CAAAA16689-03
Methyl quinoxaline-6-carboxylate 98%
Methyl quinoxaline-6-carboxylate
Formula: C₁₀H₈N₂O₂
MW: 188.19 g/mol
Melting Pt: 96…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00102618
CAS Number: 23088-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR