4-Decylbiphenyl 98%

A16685-03 A16685-06 A16685-03 A16685-06
AAA16685-06EA 344.78 USD
AAA16685-06 CAAAA16685-06 AAA16685-03 CAAAA16685-03
4-Decylbiphenyl 98%
4-Decylbiphenyl
Formula: C₂₂H₃₀
Boiling Pt: 204…210 °C/1.5 mm
Melting Pt: 52…54 °C
Storage Temperature: Ambient
MDL Number: MFCD00102117
CAS Number: 93972-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR