4-Butoxybenzonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

A16570-06 A16570-22 A16570-14 A16570-06 A16570-22 A16570-14
AAA16570-06EA 61.79 USD
AAA16570-06 AAA16570-14 CAAAA16570-06 CAAAAA16570-22 AAAA16570-22 CAAAA16570-14
4-Butoxybenzonitrile 97%
4-Butoxybenzonitrile
Formula: C₁₁H₁₃NO
Boiling Pt: 242…244 °C
Density: 0,999
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00043482
CAS Number: 5203-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR