4'-Nitrobenzo-15-crown-5 (4'-nitrobenzo-1,4,7,10,13-pentaoxacyclopentadecane) 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Nitrobenzo-15-crown-5
A16225-06
A16225-03
A16225-03
A16225-06
AAA16225-06EA
171.99
USD
CAAAA16225-03
CAAAA16225-06
AAA16225-06
AAA16225-03
4'-Nitrobenzo-15-crown-5 (4'-nitrobenzo-1,4,7,10,13-pentaoxacyclopentadecane) 99%
4'-Nitrobenzo-15-crown-5 (4'-Nitrobenzo-1,4,7,10,13-pentaoxacyclopentadecane)
Formula:
C₁₄H₁₉NO₇ MW: 313.30 g/mol Melting Pt: 93…95 °C Storage Temperature: Ambient |
MDL Number:
MFCD00060713 CAS Number: 60835-69-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...