Diacetone acrylamide 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-(1,1-Dimethyl-3-oxobutyl)acrylamide

A15940-0B A15940-0B A15940-30 A15940-18 A15940-30 A15940-18
AAA15940-18EA 50.9 USD
CAAAA15940-30 AAA15940-18 AAA15940-30 CAAAA15940-0B AAA15940-0B CAAAA15940-18
Diacetone acrylamide 99%
Diacetone acrylamide
Formula: C₉H₁₅NO₂
MW: 169.22 g/mol
Melting Pt: 54…56 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008788
CAS Number: 2873-97-4
EINECS: 220-713-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR