1-[3,5-Bis(trifluoromethyl)phenyl]pyrrole 97%

Warning

A15757-03 A15757-06 A15757-06 A15757-03
AAA15757-06EA 113.22 USD
CAAAA15757-03 AAA15757-06 AAA15757-03 CAAAA15757-06
1-[3,5-Bis(trifluoromethyl)phenyl]pyrrole 97%
1-[3,5-Bis(trifluoromethyl)phenyl]pyrrole
Formula: C₁₂H₇F₆N
MW: 279.19
Melting Pt: 41…43 °C
Storage Temperature: Ambient
MDL Number: MFCD00053048
CAS Number: 175136-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR