3-Cyanochromone 97%

Danger

Synonyms: Chromone-3-carbonitrile

A15687-03 A15687-03 A15687-06 A15687-06
AAA15687-06EA 106.1 USD
CAAAA15687-06 CAAAA15687-03 AAA15687-06 AAA15687-03
3-Cyanochromone 97%
3-Cyanochromone
Formula: C₁₀H₅NO₂
Melting Pt: 173…177 °C
Storage Temperature: Ambient
MDL Number: MFCD00052604
CAS Number: 50743-17-4
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR