4-Bromo-2-fluorobenzylamine hydrochloride 97%

Warning

Synonyms: (4-Bromo-2-fluorophenyl)methanamine hydrochloride

A15654-06 A15654-06 A15654-22 A15654-14 A15654-22 A15654-14
AAAA15654-14EA 212.28 USD
AAAA15654-14 CAAAAA15654-14 AAAA15654-22 CAAAAA15654-22 AAAA15654-06 CAAAAA15654-06
4-Bromo-2-fluorobenzylamine hydrochloride 97%
4-Bromo-2-fluorobenzylamine hydrochloride
Formula: C₇H₇BrFN·HCl
MW: 240.50 g/mol
Melting Pt: 254…257 °C
Storage Temperature: Ambient
MDL Number: MFCD00143266
CAS Number: 147181-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR