Benzofuran-2-carboxylic acid 98+%

Warning

Synonyms: Benzo[b]furan-2-carboxylic acid

A15579-03 A15579-06 A15579-14 A15579-03 A15579-06 A15579-14
AAA15579-03EA 36.47 USD
CAAAA15579-06 CAAAA15579-03 CAAAA15579-14 AAA15579-03 AAA15579-14 AAA15579-06
Benzofuran-2-carboxylic acid 98+%
Benzofuran-2-carboxylic acid
Formula: C₉H₆O₃
MW: 162.15 g/mol
Boiling Pt: 310…315 °C
Melting Pt: 192…196 °C
Storage Temperature: Ambient
MDL Number: MFCD00005848
CAS Number: 496-41-3
EINECS: 207-818-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR