Chromone-3-carboxaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Oxo-4H-chromene-3-carbaldehyde

A15438-06 A15438-14 A15438-06 A15438-22 A15438-22 A15438-14
AAA15438-06EA 35.09 USD
AAA15438-06 AAA15438-14 CAAAA15438-06 AAA15438-22 CAAAA15438-14 CAAAA15438-22
Chromone-3-carboxaldehyde 97%
Chromone-3-carboxaldehyde
Formula: C₁₀H₆O₃
MW: 174.16 g/mol
Melting Pt: 150…154 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00014667
CAS Number: 17422-74-1
EINECS: 241-451-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR